missing translation for 'onlineSavingsMsg'
Learn More

Gypsogenin, TRC

Product Code. 30008660
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008660 5 mg 5mg
30009852 1 mg 1mg
30005146 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008660 Supplier Toronto Research Chemicals Supplier No. TRCG9318085MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 639-14-5
Molecular Formula C30H46O4
Molecular Weight (g/mol) 470.68
Synonym Gypsophilasapogenin, Githagenin, Albsapogenin, Astrantiagenin D, (3β,4α)-3-Hydroxy-23-oxoolean-12-en-28-oic Acid;
IUPAC Name (4aS,6aR,6aS,6bR,8aR,9S,10S,12aR,14bS)-9-formyl-10-hydroxy-2,2,6a,6b,9,12a-hexamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
SMILES C[C@@]1(C2=O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(O)=C3CC=C)([H])[C@]4([H])CC1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C30H46O4/c1-25(2)13-15-30(24(33)34)16-14-28(5)19(20(30)17-25)7-8-22-26(3)11-10-23(32)27(4,18-31)21(26)9-12-29(22,28)6/h7,18,20-23,32H,8-17H2,1-6H3,(H,33,34)/t20-,21+,22+,23-,26-,27-,28+,29+,30-/m0/s1
Formula Weight 470.3396
Chemical Name or Material Gypsogenin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.