missing translation for 'onlineSavingsMsg'
Learn More

Gypsogenic Acid, TRC

Product Code. 30401367
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401367 25 mg -
30408455 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401367 Supplier Toronto Research Chemicals Supplier No. TRCG93180525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5143-05-5
Molecular Formula C30H46O5
Molecular Weight (g/mol) 486.68
Synonym (3β,4α)-3-Hydroxy-olean-12-ene-23,28-dioic Acid, 3β-Hydroxy-olean-12-ene-23,28-dioic Acid, Astrantiagenin J, Gypsogeninic Acid
SMILES [H][C@]12[C@](CC[C@]3(C)C2=CC[C@@]4([H])[C@@]3(C)CC[C@]5([H])[C@]4(C)CC[C@H](O)[C@@]5(C)C(O)=O)(C(O)=O)CCC(C)(C)C1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C30H46O5/c1-25(2)13-15-30(24(34)35)16-14-27(4)18(19(30)17-25)7-8-20-26(3)11-10-22(31)29(6,23(32)33)21(26)9-12-28(20,27)5/h7,19-22,31H,8-17H2,1-6H3,(H,32,33)(H,34,35)/t19-,20+,21+,22-,26+,27+,28+,29-,30-/m0/s1
Formula Weight 486.3345
Chemical Name or Material Gypsogenic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.