missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GYLTL1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563331
This item is not returnable.
View return policy
Description
Immunogen sequence: ALAQLDEEDP CFEFRQQQLT VHRVHVTFLP HEPPPPRPHD VTLVAQ Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.
Specifications
| GYLTL1B | |
| Polyclonal | |
| Unconjugated | |
| LARGE2 | |
| Beta-1,3-glucuronyltransferase LARGE2; Glycosyltransferase-like 1B; glycosyltransferase-like protein LARGE2; GYLTL1B; LARGE xylosyl- and glucuronyltransferase 2; LARGE2; like-glycosyltransferase 2; ortholog of mouse glycosyltransferase-like 1B; PP5656; Xylosyltransferase LARGE2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 120071 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N3Y3 | |
| LARGE2 | |
| Recombinant protein corresponding to Human GYLTL1B. Recombinant protein control fragment (Product #RP-101438). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction