missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GXYLT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570903
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 86%.
Glycosyltransferase which elongates the O-linked glucose attached to EGF-like repeats in the extracellular domain of Notch proteins by catalyzing the addition of xylose.
Specifications
| GXYLT1 | |
| Polyclonal | |
| Unconjugated | |
| GXYLT1 | |
| 242n2; Glt8d3; glucoside xylosyltransferase 1; glycosyltransferase 8 domain containing 3; glycosyltransferase 8 domain-containing protein 3; Glycosyltransferase 8 domain-containing protein 3-like protein; Gm1228; Gm87; GXYLT1; S33-D | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 283464 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q4G148 | |
| GXYLT1 | |
| synthetic peptide directed towards the C-terminal region of human GXYLT1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction