missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GW9508, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | 4-(((3-Phenoxyphenyl)methyl)amino)benzenepropanoic Acid |
| CAS | 885101-89-3 |
| Molecular Formula | C22H21NO3 |
| Quantity | 5 mg |
| Synonym | GW9508, 4-[[(3-Phenoxyphenyl)methyl]amino]benzenepropanoic Acid |
| InChI Formula | InChI=1S/C22H21NO3/c24-22(25)14-11-17-9-12-19(13-10-17)23-16-18-5-4-8-21(15-18)26-20-6-2-1-3-7-20/h1-10,12-13,15,23H,11,14,16H2,(H,24,25) |
| SMILES | OC(=O)CCc1ccc(NCc2cccc(Oc3ccccc3)c2)cc1 |
| IUPAC Name | 3-[4-[(3-phenoxyphenyl)methylamino]phenyl]propanoic acid |
| Molecular Weight (g/mol) | 347.41 |
| Formula Weight | 347.1521 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction