missing translation for 'onlineSavingsMsg'
Learn More

GW9508, MedChemExpress

Product Code. 16446658
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16446658

Brand: Medchem Express HY15589

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW9508 is a potent and selective G protein-coupled receptors FFA1 (GPR40) and GPR120 agonist with pEC50s of 7.32 and 5.46, respectively.

GW9508 shows ∼100-fold selectivity for GPR40 over GPR120. GW9508 is inactive against other GPCRs, kinases, proteases, integrins and PPARs. GW9508 is a glucose-sensitive insulin secretagogue and an ATP-sensitive potassium (KATP) channels opener. Anti-inflammatory and anti-atherosclerotic activities.

Specifications

Chemical Name or Material GW9508
CAS 885101-89-3
Color White
Purity Grade Notes Research
Molecular Formula C22H21NO3
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (287.84 mM)
SMILES O=C(O)CCC1=CC=C(NCC2=CC=CC(OC3=CC=CC=C3)=C2)C=C1
Molecular Weight (g/mol) 347.41
Formula Weight 347.41
Percent Purity 99.64%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.