missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GW9508, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
GW9508 shows ∼100-fold selectivity for GPR40 over GPR120. GW9508 is inactive against other GPCRs, kinases, proteases, integrins and PPARs. GW9508 is a glucose-sensitive insulin secretagogue and an ATP-sensitive potassium (KATP) channels opener. Anti-inflammatory and anti-atherosclerotic activities.
Specifications
Specifications
| Chemical Name or Material | GW9508 |
| CAS | 885101-89-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H21NO3 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (287.84 mM) |
| SMILES | O=C(O)CCC1=CC=C(NCC2=CC=CC(OC3=CC=CC=C3)=C2)C=C1 |
| Molecular Weight (g/mol) | 347.41 |
| Formula Weight | 347.41 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction