missing translation for 'onlineSavingsMsg'
Learn More

GW842166X, MedChemExpress

Product Code. 16470206
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470206 2 mg 2mg
16463406 25 mg 25mg
16455978 10 mg 10mg
16448597 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470206 Supplier Medchem Express Supplier No. HY14167

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW842166X is a potent and selective cannabinoid receptor 2 (CB2) agonist with IC50 values of 63 and 91 nM for human and rat CB2, respectively.

GW842166X is a potent and selective cannabinoid receptor 2 (CB2) agonist with IC50 values of 63 and 91 nM for human and rat CB2, respectively.

Specifications

Chemical Name or Material GW842166X
CAS 666260-75-9
Color White
Purity Grade Notes Research
Molecular Formula C18H17Cl2F3N4O2
Quantity 2 mg
Solubility Information DMSO : ≥ 83.3 mg/mL (185.42 mM)
SMILES ClC1=CC(Cl)=C(NC2=NC=C(C(NCC3CCOCC3)=O)C(C(F)(F)F)=N2)C=C1
Molecular Weight (g/mol) 449.25
Formula Weight 449.25
Percent Purity 98.73%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.