missing translation for 'onlineSavingsMsg'
Learn More

GW791343 dihydrochloride, MedChemExpress

Product Code. 16442647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442647 50 mg 50mg
16419137 5 mg 5mg
16416558 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16442647 Supplier Medchem Express Supplier No. HY15469
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GW791343 dihydrochloride is a P2X7 allosteric modulator; exhibits species-specific activity and acts as a negative allosteric modulator of human P2X7 (pIC50 = 6.9 - 7.2).

GW791343 dihydrochloride is a P2X7 allosteric modulator

Specifications

Chemical Name or Material GW791343 dihydrochloride
CAS 1019779-04-4
Color White
Purity Grade Notes Research
Molecular Formula C20H26Cl2F2N4O
Quantity 50 mg
Solubility Information H2O : 100 mg/mL (223.54 mM; Need ultrasonic) ∣DMSO : 20 mg/mL (44.71 mM; Need ultrasonic)
SMILES O=C(NC1=CC(CN2CCNCC2)=CC=C1C)CNC3=CC=C(F)C(F)=C3.[H]Cl.[H]Cl
Molecular Weight (g/mol) 447.35
Formula Weight 447.35
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.