missing translation for 'onlineSavingsMsg'
Learn More

GW1929, MedChemExpress

Product Code. 16453486
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16453486 25 mg 25mg
16432807 50 mg 50mg
16429277 5 mg 5mg
16416728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16453486 Supplier Medchem Express Supplier No. HY15655
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GW1929 is a potent PPAR-γ agonist, with a pKi of 8.84 for human PPAR-γ, and pEC50s of 8.56 and 8.27 for human PPAR-γ and murine PPAR-γ, respectively.

GW1929 is a potent PPAR-γ agonist, with a pKi of 8.84 for human PPAR-γ, and pEC50s of 8.56 and 8.27 for human PPAR-γ and murine PPAR-γ, respectively.

Specifications

Chemical Name or Material GW1929
CAS 196808-24-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H29N3O4
Quantity 25 mg
Solubility Information DMSO : ≥ 35 mg/mL (70.63 mM)
SMILES O=C(O)[C@H](CC1=CC=C(C=C1)OCCN(C)C2=NC=CC=C2)NC3=CC=CC=C3C(C4=CC=CC=C4)=O
Molecular Weight (g/mol) 495.57
Formula Weight 495.57
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.