missing translation for 'onlineSavingsMsg'
Learn More

GW182 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18394525 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394525 20 μL 20µL
18304046 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18394525 Supplier Novus Biologicals Supplier No. NBP30301420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GW182 Polyclonal specifically detects GW182 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GW182
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias CAG repeat protein 26, CAGH26FLJ22043, DKFZp666E117, EMSY interactor protein, Glycine-tryptophan protein of 182 kDa, GW182 autoantigen, GW182GW1, KIAA1460EDIE, MGC75384, Protein GW1, TNRC6, trinucleotide repeat containing 6, trinucleotide repeat containing 6A, trinucleotide repeat-containing gene 6A protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 950-1050 of human GW182 (NP_055309.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27327
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.