missing translation for 'onlineSavingsMsg'
Learn More

GW 9662, Thermo Scientific Chemicals

Product Code. 15486999
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486999 10 mg 10mg
15496999 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15486999 Supplier Thermo Scientific Alfa Aesar Supplier No. J65634.MA
  • ONLINE EXCLUSIVE PRICE
    £89.40
    £80.65 / 10mg
    Save £8.75
    9% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An irreversible PPAR→ antagonist

An irreversible PPAR- antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22978-25-2
Molecular Formula C13H9ClN2O3
Molecular Weight (g/mol) 276.676
MDL Number MFCD01215270
InChI Key DNTSIBUQMRRYIU-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzanilide, 2-chloro-5-nitro-n-phenyl-benzamide, 2-chloro-5-nitro-n-4-phenylbenzamide, 2-chloro-5-nitrophenyl-n-benzamide, tocris-1508, spectrum5_001989, lopac-m-6191, d00acx, d05thb, dsstox_cid_20723
PubChem CID 644213
ChEBI CHEBI:79993
IUPAC Name 2-chloro-5-nitro-N-phenylbenzamide
SMILES C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl

Specifications

Color White
Quantity 10 mg
Formula Weight 276.68
Physical Form Powder
Chemical Name or Material GW 9662
compliance-icons
Product Identifier
  • GW 9662
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Chloro-5-nitrobenzanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.