missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GW 791343 hydrochloride, Tocris Bioscience™
P2X7 allosteric modulator
Chemical Identifiers
| CAS | 309712-55-8 |
|---|---|
| Molecular Formula | C20H27Cl3F2N4O |
| Molecular Weight (g/mol) | 483.81 |
| InChI Key | WSBRAHWNJBXXJM-UHFFFAOYSA-N |
| Synonym | gw 791343 hydrochloride, gw791343 trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-1-piperazinylmethyl phenyl-acetamide trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-piperazin-1-ylmethyl phenyl acetamide trihydrochloride, d05exn, gw-791343 hydrochloride, gw 791343 trihydrochloride, c20h24f2n4o.3clh, gw 791343 trihydrochloride;gw-791343 trihydrochloride |
| PubChem CID | 9848159 |
| IUPAC Name | 2-(3,4-difluoroanilino)-N-[2-methyl-5-(piperazin-1-ylmethyl)phenyl]acetamide;trihydrochloride |
| SMILES | CC1=C(C=C(C=C1)CN2CCNCC2)NC(=O)CNC3=CC(=C(C=C3)F)F.Cl.Cl.Cl |
Description
P2X7 allosteric modulator. Exhibits species-specific activity; acts as a negative allosteric modulator of human P2X7 (pIC50 = 6.9 - 7.2) and a positive allosteric modulator of rat P2X7.Chemical Identifiers
| 309712-55-8 | |
| 483.81 | |
| gw 791343 hydrochloride, gw791343 trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-1-piperazinylmethyl phenyl-acetamide trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-piperazin-1-ylmethyl phenyl acetamide trihydrochloride, d05exn, gw-791343 hydrochloride, gw 791343 trihydrochloride, c20h24f2n4o.3clh, gw 791343 trihydrochloride;gw-791343 trihydrochloride | |
| 2-(3,4-difluoroanilino)-N-[2-methyl-5-(piperazin-1-ylmethyl)phenyl]acetamide;trihydrochloride |
| C20H27Cl3F2N4O | |
| WSBRAHWNJBXXJM-UHFFFAOYSA-N | |
| 9848159 | |
| CC1=C(C=C(C=C1)CN2CCNCC2)NC(=O)CNC3=CC(=C(C=C3)F)F.Cl.Cl.Cl |
Specifications
| 309712-55-8 | |
| gw 791343 hydrochloride, gw791343 trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-1-piperazinylmethyl phenyl-acetamide trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-piperazin-1-ylmethyl phenyl acetamide trihydrochloride, d05exn, gw-791343 hydrochloride, gw 791343 trihydrochloride, c20h24f2n4o.3clh, gw 791343 trihydrochloride;gw-791343 trihydrochloride | |
| CC1=C(C=C(C=C1)CN2CCNCC2)NC(=O)CNC3=CC(=C(C=C3)F)F.Cl.Cl.Cl | |
| 483.81 | |
| 483.81 | |
| GW 791343 hydrochloride |
| C20H27Cl3F2N4O | |
| WSBRAHWNJBXXJM-UHFFFAOYSA-N | |
| 2-(3,4-difluoroanilino)-N-[2-methyl-5-(piperazin-1-ylmethyl)phenyl]acetamide;trihydrochloride | |
| 9848159 | |
| >99% |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.