missing translation for 'onlineSavingsMsg'
Learn More

GW 4064, MedChemExpress

Product Code. 16440528
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16440528

Brand: Medchem Express HY50108

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW 4064 is a potent FXR agonist with an EC50 of 65 nM.

GW 4064 is a potent FXR agonist with an EC50 of 65 nM.

Specifications

Chemical Name or Material GW 4064
CAS 278779-30-9
Color White
Purity Grade Notes Research
Molecular Formula C28H22Cl3NO4
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (230.27 mM; Need ultrasonic) ∣DMF : 100 mg/mL (184.22 mM; Need ultrasonic)
SMILES O=C(C1=CC=CC(/C=C/C2=CC=C(C=C2Cl)OCC3=C(ON=C3C4=C(Cl)C=CC=C4Cl)C(C)C)=C1)O
Molecular Weight (g/mol) 542.84
Formula Weight 542.84
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.