missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GW 3965 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | GW 3965 Hydrochloride |
| CAS | 405911-17-3 |
| Molecular Formula | C33H31ClF3NO3 •HCl |
| Quantity | 100 mg |
| Synonym | GW 3965A Hydrochloride, 2-(3-(3-((2-Chloro-3-(trifluoromethyl)benzyl)(2,2-diphenylethyl)amino)propoxy)phenyl)acetic Acid Hydrochloride, 3-[3-[N-(2-Chloro-3-trifluoromethylbenzyl)(2,2-diphenylethyl)amino]propoxy]phenylacetic Acid Hydrochloride, 3-[3-[[[2-Chloro-3-(trifluoromethyl)phenyl]methyl](2,2-diphenylethyl)amino]propoxy]-benzeneacetic Acid Hydrochloride |
| InChI Formula | InChI=1S/C33H31ClF3NO3.ClH/c34-32-27(15-8-17-30(32)33(35,36)37)22-38(18-9-19-41-28-16-7-10-24(20-28)21-31(39)40)23-29(25-11-3-1-4-12-25)26-13-5-2-6-14-26;/h1-8,10-17,20,29H,9,18-19,21-23H2,(H,39,40);1H |
| SMILES | ClC(C(C(F)(F)F)=CC=C1)=C1CN(CC(C2=CC=CC=C2)C3=CC=CC=C3)CCCOC4=CC(CC(O)=O)=CC=C4.Cl |
| Molecular Weight (g/mol) | 582.053646 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?