missing translation for 'onlineSavingsMsg'
Learn More

GW 3965 Hydrochloride, TRC

Product Code. 30393844
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393844 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393844 Supplier Toronto Research Chemicals Supplier No. TRCG93098510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material GW 3965 Hydrochloride
CAS 405911-17-3
Molecular Formula C33H31ClF3NO3 •HCl
Quantity 10 mg
Synonym GW 3965A Hydrochloride, 2-(3-(3-((2-Chloro-3-(trifluoromethyl)benzyl)(2,2-diphenylethyl)amino)propoxy)phenyl)acetic Acid Hydrochloride, 3-[3-[N-(2-Chloro-3-trifluoromethylbenzyl)(2,2-diphenylethyl)amino]propoxy]phenylacetic Acid Hydrochloride, 3-[3-[[[2-Chloro-3-(trifluoromethyl)phenyl]methyl](2,2-diphenylethyl)amino]propoxy]-benzeneacetic Acid Hydrochloride
InChI Formula InChI=1S/C33H31ClF3NO3.ClH/c34-32-27(15-8-17-30(32)33(35,36)37)22-38(18-9-19-41-28-16-7-10-24(20-28)21-31(39)40)23-29(25-11-3-1-4-12-25)26-13-5-2-6-14-26;/h1-8,10-17,20,29H,9,18-19,21-23H2,(H,39,40);1H
SMILES ClC(C(C(F)(F)F)=CC=C1)=C1CN(CC(C2=CC=CC=C2)C3=CC=CC=C3)CCCOC4=CC(CC(O)=O)=CC=C4.Cl
Molecular Weight (g/mol) 582.053646
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.