missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GW 280264X

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GW 280264X is a potent ADAM (A Disintegrin and Metalloproteinase) 17 and ADAM10 inhibitor (IC50 values of 8 nM and 11.5 nM, respectively). It inhibits constitutive and PMA-inducible CX3CL1 shedding and increases adhesive properties in CX3CL1 transfected cells (IC50 = 1 μM in COS-7 and ECV-304 cells). GW 280264X treatment in mice significantly improves functional recovery after spinal cord injury.
Specifications
Specifications
| Chemical Name or Material | Phenylmethyl N-[(5 S)-5-[[(2 R,3 S)-3-(formylhydroxyamino)-2-(2-methylpropyl)-1-oxohexyl]amino]-6-oxo-6-(2-thiazolylamino)hexyl]carbamate |
| CAS | 866924-39-2 |
| Quantity | 10 mg |
| Target | Synthetic Metalloprotease Inhibitors |
| Molecular Formula | C28H41N5O6S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction