missing translation for 'onlineSavingsMsg'
Learn More

GW-1100, MedChemExpress

Product Code. p-200099810
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16470548

Brand: Medchem Express HY50691

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW-1100 is a selective GPR40 antagonist with a pIC50 of 6.9.

GW-1100 is a selective GPR40 antagonist with a pIC50 of 6.9.

Specifications

Chemical Name or Material GW-1100
CAS 306974-70-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H25FN4O4S
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (96.05 mM; Need ultrasonic)
SMILES O=C1C(CC2=CN=C(N=C2)OCC)=CN(C(SCC3=CC=C(C=C3)F)=N1)C4=CC=C(C(OCC)=O)C=C4
Molecular Weight (g/mol) 520.58
Formula Weight 520.58
Percent Purity 97.01%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.