missing translation for 'onlineSavingsMsg'
Learn More

Guvacoline Hydrochloride, TRC

Product Code. 30400863
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400863 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400863 Supplier Toronto Research Chemicals Supplier No. TRCG87651025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Guvacoline Hydrochloride
CAS 6197-39-3
Molecular Formula C7H11NO2 HCl
Quantity 25 mg
Synonym Methyl 1,2,5,6-Tetrahydro-3-pyridinecarboxylate Hydrochloride
InChI Formula InChI=1S/C7H11NO2.ClH/c1-10-7(9)6-3-2-4-8-5-6;/h3,8H,2,4-5H2,1H3;1H
SMILES Cl.COC(=O)C1=CCCNC1
IUPAC Name methyl 1,2,3,6-tetrahydropyridine-5-carboxylate;hydrochloride
Molecular Weight (g/mol) 177.63
Formula Weight 177.0557
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.