missing translation for 'onlineSavingsMsg'
Learn More

Guvacine hydrochloride, MedChemExpress

Product Code. 16423178
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16423178

Brand: Medchem Express HY100809

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Guvacine hydrochloride is an alkaloid from the nut of Areca catechu, acts as an inhibitor of GABA transporter, and dispalys modest selectivity for cloned GABA transporters with IC50s of 14 μM (human GAT-1), 39 μM (rat GAT-1), 58 μM (rat GAT-2), 119 μM (human GAT-3), 378 μM (rat GAT-3), and 1870 μM (human BGT-3).

Guvacine hydrochloride is an alkaloid from the nut of Areca catechu, acts as an inhibitor of GABA transporter, and dispalys modest selectivity for cloned GABA transporters with IC50s of 14 μM (human GAT-1), 39 μM (rat GAT-1), 58 μM (rat GAT-2), 119 μM (human GAT-3), 378 μM (rat GAT-3), and 1870 μM (human BGT-3).

Specifications

Chemical Name or Material Guvacine hydrochloride
CAS 6027-91-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C6H10ClNO2
Quantity 10 mg
Solubility Information H2O : 41.67 mg/mL (254.71 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (6.11 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(C1=CCCNC1)O.[H]Cl
Molecular Weight (g/mol) 163.6
Formula Weight 163.6
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.