missing translation for 'onlineSavingsMsg'
Learn More

Guthoxon, TRC

Product Code. 30399752
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399752 100 mg 100mg
30404169 50 mg -
30390718 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399752 Supplier Toronto Research Chemicals Supplier No. TRCG855650100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 961-22-8
Molecular Formula C10 H12 N3 O4 P S
Molecular Weight (g/mol) 301.26
Synonym Phosphorothioic acid, O,O-dimethyl ester, S-ester with 3-(mercaptomethyl)-1,2,3-benzotriazin-4(3H)-one (6CI,7CI,8CI), 1,2,3-Benzotriazine, phosphorothioic acid deriv., Azinphosmethyl oxon, Azinphosmethyl oxygen analog, Guthion oxon, Guthion oxygen analog, Guthoxon, Gutoxon, O,O-Dimethyl S-(4-oxobenzotriazino-3-methyl) phosphorothiolate, Oxoazinphos-methyl, S-3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-ylmethyl O,O-dimethyl phosphorothioate, Phosphorothioic acid, O,O-dimethyl S-[(4-oxo-1,2,3-benzotriazin-3(4H)-yl)methyl] ester
IUPAC Name 3-(dimethoxyphosphorylsulfanylmethyl)-1,2,3-benzotriazin-4-one
SMILES COP(=O)(OC)SCN1N=Nc2ccccc2C1=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H12N3O4PS/c1-16-18(15,17-2)19-7-13-10(14)8-5-3-4-6-9(8)11-12-13/h3-6H,7H2,1-2H3
Formula Weight 301.0286
Chemical Name or Material Azinphos-methyl-oxon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.