missing translation for 'onlineSavingsMsg'
Learn More

Gum guaiac, MP Biomedicals™

Product Code. 11423792 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 11423792

Brand: MP Biomedicals 0215729191

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 9000-29-7

Specifications

Color Brown to Brown/Green
Quantity 1 kg
Physical Form Powder
Chemical Name or Material Gum guaiac
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.