missing translation for 'onlineSavingsMsg'
Learn More

Gum arabic, MP Biomedicals™

Product Code. 11483751 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483751 1 kg 1kg
11463751 100 g 100g
11473751 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483751 Supplier MP Biomedicals Supplier No. 0215121891
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gum Arabic can be branched polymer of galactose, rhamnose, arabinose, and glucuronic acid as the calcium, magnesium, and potassium salts.

  • It is used in textiles, ceramics, lithography, cosmetics and pharmaceuticals.
  • In the food industry, acacia is mainly used as confectionery, bakery, dairy, drink, and as a microencapsulating agent.

Chemical Identifiers

CAS 9000-01-5

Specifications

Color White
Physical Form Powder
Synonym Arabic gum, Australian gum
Quantity 1 kg
Formula Weight 250,000 to 1,000,000
Chemical Name or Material Gum arabic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.