missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GULP1 Polyclonal Antibody

Description
Recommended positive controls: 293T, HepG2, Molt-4, NIH-3T3. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The prompt clearance of cells undergoing apoptosis is critical during embryonic development, normal tissue turnover, inflammation, and autoimmunity. CED6 is an evolutionarily conserved adaptor protein required for efficient engulfment of apoptotic cells by phagocytes.
Specifications
Specifications
| Antigen | GULP1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | GULP1 |
| Gene Accession No. | Q5PQS4, Q8K2A1, Q9UBP9 |
| Gene Alias | 3110030A04Rik; 5730529O06Rik; CED 6; Ced6; CED-6; CED-6 protein; Cell death protein 6 homolog; engulfment adapter protein; FLJ31156; GULP; GULP, engulfment adaptor PTB domain containing 1; GULP1; Gulp-2; OTTHUMP00000205908; OTTHUMP00000205909; OTTHUMP00000205911; Protein GULP; PTB domain adapter protein CED-6; PTB domain adaptor protein CED-6; PTB domain-containing engulfment adapter protein 1 |
| Gene Symbols | GULP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?