missing translation for 'onlineSavingsMsg'
Learn More

GULP1/CED-6 Antibody [PerCP], Novus Biologicals™

Product Code. 30737493 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737493 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737493 Supplier Novus Biologicals Supplier No. AF7978C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

GULP1/CED-6 Polyclonal antibody specifically detects GULP1/CED-6 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GULP1/CED-6
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias CED6GULPCED-6, Cell death protein 6 homolog, engulfment adapter protein, FLJ31156, GULP, engulfment adaptor PTB domain containing 1, Protein GULP, PTB domain adapter protein CED-6, PTB domain adaptor protein CED-6, PTB domain-containing engulfment adapter protein 1
Host Species Sheep
Immunogen E. coli-derived recombinant human GULP1/CED-6, Met1-Asn92, Accession # Q9UBP9
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 51454
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.