missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Guinea Pig Mineral Mix

Product Code. 11497221 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497221 2 kg 2kg
11407231 10 kg 10kg
11417231 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497221 Supplier MP Biomedicals™ Supplier No. 0296028502
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides complete minerals for normal growth and development of the guinea pig when added at a level of 6.75% of the diet.

  • Salt Mixture
  • For guinea pig diet

Specifications

Product Type Animal Diet
For Use With (Application) Guinea pig diet
Quantity 2 kg
Storage Requirements Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.