missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guinea Pig anti-Rabbit IgG (H+L) Secondary Antibody, DyLight 550 (Pre-adsorbed), Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Host Species | Guinea Pig |
| Immunogen | Rabbit IgG whole molecule |
| Purification Method | Multi-step |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?