missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYY, Guinea pig, Polyclonal Antibody, Abnova™
Guinea pig polyclonal antibody raised against synthetic peptide of PYY.
Brand: Abnova PAB13801.50uL
This item is not returnable.
View return policy
Specifications
| PYY | |
| Polyclonal | |
| Guinea pig polyclonal antibody raised against synthetic peptide of PYY. | |
| Lyophilized from antiserum | |
| PYY | |
| A synthetic peptide corresponding to pig PYY. | |
| RUO | |
| 445018 | |
| Store at 4°C on dry atmosphere. After reconstitution with distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1500-1:2500) Immunohistochemistry (Frozen sections) (1:1500-1:2500) The optimal working dilution should be determined by the end user. | |
| PYY | |
| Guinea Pig | |
| 50 μL | |
| Primary | |
| Feline, Mouse, Pig, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction