Learn More
OXT, Guinea pig, Polyclonal Antibody, Abnova™
Guinea pig polyclonal antibody raised against synthetic peptide of OXT.
Brand: Abnova PAB14310.50uL
Description
There are two proteins encoded by this gene, oxytocin and neurophysin I. Oxytocin is posterior pituitary hormone which is synthesized as an inactive precursor in the hypothalamus along with its carrier protein neurophysin I. Together with neurophysin, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis, where it is either stored or secreted into the bloodstream. The precursor seems to be activated while it is being transported along the axon to the posterior pituitary. This hormone contracts smooth muscle during parturition and lactation. It is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions. [provided by RefSeq
Sequence: YIQNCPLGSpecifications
| OXT | |
| Polyclonal | |
| Guinea pig polyclonal antibody raised against synthetic peptide of OXT. | |
| Lyophilized from PBS | |
| MGC126890/MGC126892/OT/OT-NPI | |
| Guinea Pig | |
| 50 μL | |
| Primary | |
| Human, Sheep | |
| Lyophilized |
| Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Immunohistochemistry (1:2000) The optimal working dilution should be determined by the end user. | |
| OXT | |
| OXT | |
| A synthetic peptide (conjugated with carrier protein) corresponding to amino acids 20-28 of human OXT. | |
| RUO | |
| 5020 | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only