missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GUCY2F Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04892-100UL
This item is not returnable.
View return policy
Description
GUCY2F Polyclonal specifically detects GUCY2F in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| GUCY2F | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| CYGF, EC 4.6.1, GC-FEC 4.6.1.2, guanylate cyclase 2F, retinalRETGC-2, Guanylate cyclase F, GUC2DL, GUC2Fguanylate cyclase 2D-like, membrane (retina-specific), RETGC2, retinal guanylyl cyclase 2, Rod outer segment membrane guanylate cyclase 2, ROS-GC2RetGC-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-460 of human GUCY2F (NP_001513.2). | |
| 100 μL | |
| Protein Kinase | |
| 2986 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction