missing translation for 'onlineSavingsMsg'
Learn More

GUCY2D Antibody (X1), Novus Biologicals™

Product Code. 18328517 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328517 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328517 Supplier Novus Biologicals Supplier No. H00003000M09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GUCY2D Monoclonal antibody specifically detects GUCY2D in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GUCY2D
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone X1
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000171
Gene Alias CORD6cone rod dystrophy 5/6, CYGD, EC 4.6.1, guanylate cyclase 2D, membrane (retina-specific), Guanylate cyclase 2D, retinal, GUC1A4, GUC2DCORD5, LCA, LCA1RCD2, retGC, RETGC1, RETGC-1EC 4.6.1.2, retinal guanylyl cyclase 1, Rod outer segment membrane guanylate cyclase, ROSGC, ROS-GC, ROS-GC1
Host Species Mouse
Immunogen GUCY2D (NP_000171, 521 a.a. ~ 630 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RKVAQGSRSSLGARSMSDIRSGPSQHLDSPNIGVYEGDRVWLKKFPGDQHIAIRPATKTAFSKLQELRHENVALYLGLFLARGAEGPAALWEGNLAVVSEHCTRGSLQDL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3000
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.