missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GUCY2D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35345-20ul
This item is not returnable.
View return policy
Description
GUCY2D Polyclonal antibody specifically detects GUCY2D in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| GUCY2D | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| CORD6cone rod dystrophy 5/6, CYGD, EC 4.6.1, guanylate cyclase 2D, membrane (retina-specific), Guanylate cyclase 2D, retinal, GUC1A4, GUC2DCORD5, LCA, LCA1RCD2, retGC, RETGC1, RETGC-1EC 4.6.1.2, retinal guanylyl cyclase 1, Rod outer segment membrane guanylate cyclase, ROSGC, ROS-GC, ROS-GC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 52-350 of human GUCY2D (NP_000171.1).,, Sequence:, AVFTVGVLGPWACDPIFSRARPDLAARLAAARLNRDPGLAGGPRFEVALLPEPCRTPGSLGAVSSALARVSGLVGPVNPAACRPAELLAEEAGIALVPWGCPWTQAEGTTAPAVTPAADALYALLRAFGWARVALVTAPQDLWVEAGRSLSTALRARGLPVASVTSMEPLDLSGAREALRKVRDGPRVTAVIMVMHSVLLGGEEQRYLLEAAEELGLTDGSLVFLPFDTIHYALSPGPEALAALANSSQLRRAHDAVLTLTRHCPSEGSVLDSLRRAQERRELPSDLNLQQVSPLFGTI | |
| 20 μL | |
| Protein Kinase | |
| 3000 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion