missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GUCY1B2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103135
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GCS beta 2.
Guanylate cyclases belong to the adenylyl cyclase class-4/guanylyl cyclase family. There are two forms of guanylate cyclase. The soluble form, known as GCS or sGC, act as receptors for nitric oxide. The membrane-bound receptor form, known as GC, are peptide hormone receptors. GCS is a cGMP-synthesizing enzyme, which is the major receptor for the neurotransmitter nitric oxide (NO). It plays a crucial role in smooth muscle contractility, platelet reactivity and neurotransmission. GCS is a heme containing heterodimer, consisting of one 72 kDa alpha subunit and one 65 kDa beta subunit. The heme moeity mediates NO activation, and this heme group also binds carbon monoxide, which weakly stimulates the enzyme.
Specifications
| GUCY1B2 | |
| Polyclonal | |
| Unconjugated | |
| GUCY1B2 | |
| B930025B03; GCS beta2; GC-SB2; GCS-beta2; GCS-beta-2; guanylate cyclase 1 soluble subunit beta 2 (pseudogene); guanylate cyclase 1, soluble, beta 2; Guanylate cyclase soluble beta 2 (GTP pyrophosphate - lyase); guanylate cyclase soluble subunit beta-2; Guanylate cyclase, soluble, beta 2 (GTP pyrophosphate - lyase); Gucy1b2; Gucy1b2a; Gucy1b2b; SGC; soluble guanylate cyclase beta 2a; soluble guanylate cyclase beta 2b; soluble guanylyl cyclase beta2 subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 239134, 25206, 2974 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75343, P22717 | |
| GUCY1B2 | |
| A synthesized peptide derived from human GUCY1B2(Accession O75343), corresponding to amino acid residues P581-L617. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction