missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GUCY1A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115335
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Guanylyl Cyclase alpha 2.
Soluble guanylate cyclase (sGC), a heterodimeric protein consisting of an alpha and a beta subunit, catalyzes the conversion of GTP to the second messenger cGMP and functions as the main receptor for nitric oxide and nitrovasodilator drugs.
Specifications
| GUCY1A2 | |
| Polyclonal | |
| Unconjugated | |
| GUCY1A2 | |
| 6330407I18Rik; A230060L24Rik; GC-SA2; GCS-alpha-2; guanylate cyclase 1 soluble subunit alpha 2; guanylate cyclase 1, soluble, alpha 2; guanylate cyclase soluble subunit alpha-2; GUC1A2; GUCSA2; Gucy1a2; soluble guanylyl cyclase alpha2 subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 234889, 2977, 66012 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P33402, Q9WVI4 | |
| GUCY1A2 | |
| A synthesized peptide derived from human GUCY1A2(Accession P33402), corresponding to amino acid residues H118-E168. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction