missing translation for 'onlineSavingsMsg'
Learn More

Guanylyl Imidodiphosphate Trisodium Salt (90%), TRC

Product Code. 30393945
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393945 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393945 Supplier Toronto Research Chemicals Supplier No. TRCG8447001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Guanylyl Imidodiphosphate Trisodium Salt
CAS 148892-91-5
Molecular Formula C10H14N6Na3O13P3
Quantity 1 mg
Synonym 5'-Guanylic Acid Monoanhydride with Imidodiphosphoric Acid Trisodium Salt
InChI Formula InChI=1S/C10H17N6O13P3.3Na/c11-10-13-7-4(8(19)14-10)12-2-16(7)9-6(18)5(17)3(28-9)1-27-32(25,26)29-31(23,24)15-30(20,21)22;;;/h2-3,5-6,9,17-18H,1H2,(H,25,26)(H3,11,13,14,19)(H4,15,20,21,22,23,24);;;/q;3*+1/p-3/t3-,5-,6-,9-;;;/m1.../s1
SMILES [Na+].[Na+].[Na+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(=O)([O-])OP(=O)([O-])NP(=O)(O)[O-])[C@@H](O)[C@H]3O)C(=O)N1
IUPAC Name trisodium;[[(2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl]oxy-[[hydroxy(oxido)phosphoryl]amino]phosphinate
Molecular Weight (g/mol) 588.14
Formula Weight 587.9525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.