missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanylyl Cyclase alpha 1 Antibody (SR2180), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22388-50ul
This item is not returnable.
View return policy
Description
Guanylyl Cyclase alpha 1 Monoclonal antibody specifically detects Guanylyl Cyclase alpha 1 in Human samples. It is validated for Western Blot
Specifications
| Guanylyl Cyclase alpha 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR2180 | |
| Western Blot 1:500-1:2000 | |
| EC 4.6.1.2, GC-S-alpha-1, GCS-alpha-1, GCS-alpha-3, guanylate cyclase 1, soluble, alpha 3, guanylate cyclase soluble subunit alpha-3, GUC1A3GUCA3, GUCSA3GC-SA3, GUCY1A1, Soluble guanylate cyclase large subunit | |
| A synthesized peptide derived from human Guanylyl Cyclase alpha 1 (Uniprot #: Q02108) | |
| 50 μg | |
| Signal Transduction, Vision | |
| 2982 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction