missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanylyl Cyclase alpha 1 Antibody (3G6B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£184.00 - £449.00
Specifications
| Antigen | Guanylyl Cyclase alpha 1 |
|---|---|
| Clone | 3G6B2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18151789
|
Novus Biologicals
NBP2-37401 |
0.1 mL |
£449.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628605
|
Novus Biologicals
NBP2-37401-0.025ml |
0.025 mL |
£184.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Guanylyl Cyclase alpha 1 Monoclonal specifically detects Guanylyl Cyclase alpha 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Guanylyl Cyclase alpha 1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 4.6.1.2, GC-S-alpha-1, GCS-alpha-1, GCS-alpha-3, guanylate cyclase 1, soluble, alpha 3, guanylate cyclase soluble subunit alpha-3, GUC1A3GUCA3, GUCSA3GC-SA3, GUCY1A1, Soluble guanylate cyclase large subunit | |
| GUCY1A1 | |
| IgG1 | |
| Affinity Purified |
| 3G6B2 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Signal Transduction, Vision | |
| PBS with 0.05% Sodium Azide | |
| 2982 | |
| Purified recombinant fragment of human GUCY1A3 (AA: 22-214) expressed in E. Coli. | |
| Primary | |
| 77.5 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title