missing translation for 'onlineSavingsMsg'
Learn More

Guanyl Urea Sulfate, TRC

Product Code. 30397683
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397683 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397683 Supplier Toronto Research Chemicals Supplier No. TRCG844500100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Guanylurea Sulfate
CAS 591-01-5
Molecular Formula 2 C2 H6 N4 O . H2 O4 S
Quantity 100 g
Synonym Urea, N-(aminoiminomethyl)-, sulfate (2:1), Urea, (aminoiminomethyl)-, sulfate (2:1) (9CI), Urea, amidino-, sulfate (2:1) (8CI), Carbamylguanidine sulfate, Dicyanodiamidine sulfate, Diuretamidine sulfate, Guanylurea sulfate, N-Guanylurea Sulfate
InChI Formula InChI=1S/2C2H6N4O.H2O4S/c2*3-1(4)6-2(5)7;1-5(2,3)4/h2*(H6,3,4,5,6,7);(H2,1,2,3,4)
SMILES NC(=N)NC(=O)N.NC(=N)NC(=O)N.OS(=O)(=O)O
IUPAC Name carbamimidoylurea;sulfuric acid
Molecular Weight (g/mol) 302.27
Formula Weight 302.0757
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.