missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanosine-5'-triphosphate Trisodium Salt, TRC
Click to view available options
Quantity:
25 mg
1 g
Unit Size:
1g
25mg
Specifications
Specifications
| Chemical Name or Material | Guanosine-5'-triphosphate Trisodium Salt |
| CAS | 36051-31-7 |
| Molecular Formula | C10H13N5Na3O14P3 |
| Quantity | 1 g |
| Synonym | Guanosine 5'-(tetrahydrogen Triphosphate), Trisodium Salt, 5'-GTP Di-Na Salt, Trisodium 5'-GTP, GTP Trisodium Salt |
| InChI Formula | InChI=1S/C10H16N5O14P3.3Na/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(27-9)1-26-31(22,23)29-32(24,25)28-30(19,20)21;;;/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H,24,25)(H2,19,20,21)(H3,11,13,14,18);;;/q;3*+1/p-3/t3-,5-,6-,9-;;;/m1.../s1 |
| SMILES | [Na+].[Na+].[Na+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(=O)([O-])OP(=O)([O-])OP(=O)(O)[O-])[C@@H](O)[C@H]3O)C(=O)N1 |
| IUPAC Name | trisodium;[[[(2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl]oxy-oxidophosphoryl] hydrogen phosphate |
| Molecular Weight (g/mol) | 588.94 |
| Formula Weight | 588.9365 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction