missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Guanosine 5'-triphosphate, disodium salt hydrate, 90%, for biochemistry

Product Code. 10012880 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10012880 250 mg Glass bottle 250mg
10184750 50 mg Glass bottle 50mg
10732461 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10012880 Supplier Thermo Scientific Chemicals Supplier No. 226252500
  • ONLINE EXCLUSIVE PRICE
    £197.00
    £170.65 / 250mg
    Save £26.35
    13% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Guanosine 5'-triphosphate, disodium salt hydrate, 90%, C10H14N5Na2O14P3.xH2O, CAS Number-56001-37-7 | CAS: 56001-37-7 | C10H14N5Na2O14P3 | 567.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56001-37-7
Molecular Formula C10H14N5Na2O14P3
Molecular Weight (g/mol) 567.14
MDL Number MFCD03410297
InChI Key FIZIYLKEXVIRHJ-KHRSEZDTNA-L
Synonym guanosine 5'-triphosphate, guanosine-5'-triphosphate disodium salt dihydrate gtp, guanosine-triphosphate, guanosine 5'-triphosphate 4-, gtp 4-, 2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate, 2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate
PubChem CID 131676145
IUPAC Name disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate
SMILES [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Absorbance 1.12 to 1.18 (A250/A260), 0.63 to 0.69 (A280/A260)
Melting Point 180°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: soluble
Spectrophotometry 88% min. (lambda max. 252nm,pH 7)
Formula Weight 567.14
Percent Purity 90%
Related Substances 4% max. (typically) Guanosine diphosphate (HPLC)
Grade Biochemistry
Physical Form Crystalline Powder
Chemical Name or Material Guanosine 5'-triphosphate, disodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.