missing translation for 'onlineSavingsMsg'
Learn More

Guanosine-5'-triphosphate Disodium Salt (~85%), TRC

Product Code. 30399322
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30399322 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399322 Supplier Toronto Research Chemicals Supplier No. TRCG838608500MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Guanosine-5'-triphosphate Disodium Salt
CAS 56001-37-7
Molecular Formula C10H14N5O14P3Na2
Quantity 500 mg
Synonym Guanosine 5'-(tetrahydrogen triphosphate) sodium salt (1:2), Guanosine 5'-(tetrahydrogen triphosphate) disodium salt (9CI), 5’-GTP di-Na salt, Disodium 5'-GTP, Disodium guanosine triphosphate, GTP disodium salt, Guanosine 5'-triphosphoric acid disodium salt
InChI Formula InChI=1S/C10H16N5O14P3.2Na/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(27-9)1-26-31(22,23)29-32(24,25)28-30(19,20)21;;/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H,24,25)(H2,19,20,21)(H3,11,13,14,18);;/q;2*+1/p-2/t3-,5-,6-,9-;;/m1../s1
SMILES [Na+].[Na+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(=O)(O)OP(=O)(O)OP(=O)([O-])[O-])[C@@H](O)[C@H]3O)C(=O)N1
IUPAC Name disodium;[[[(2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate
Molecular Weight (g/mol) 567.14
Formula Weight 566.9545
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.