missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanosine 5'-Pyrophosphate Ester 6-Deoxytalose (>90%) Disodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Guanosine 5'-Pyrophosphate Ester 6-Deoxytalose Disodium Salt |
| Molecular Formula | C16 H23 N5 Na2 O15 P2 |
| Quantity | 0.25 mg |
| Synonym | P'-(6-Deoxy-alpha-D-talopyranosyl) Ester Guanosine 5'-(Trihydrogen Diphosphate) Disodium Salt, Mono(6-deoxy-alpha-D-talopyranosyl) Ester Guanosine 5'-(Trihydrogen Pyrophosphate) Disodium Salt, Guanosine 5'-Pyrophosphate, Ester alpha-D-6-Deoxytalopyranosyl Disodium Salt, 6-Deoxytalopyranose 1-Ester Guanosine 5’-(Trihydrogen Pyrophosphate) Disodium Salt |
| InChI Formula | InChI=1S/C16H25N5O15P2.2Na/c1-4-7(22)9(24)11(26)15(33-4)35-38(30,31)36-37(28,29)32-2-5-8(23)10(25)14(34-5)21-3-18-6-12(21)19-16(17)20-13(6)27;;/h3-5,7-11,14-15,22-26H,2H2,1H3,(H,28,29)(H,30,31)(H3,17,19,20,27);;/q;2*+1/p-2/t4-,5-,7+,8-,9?,10-,11+,14-,15+;;/m1../s1 |
| SMILES | C[C@H]1O[C@@H](OP(=O)(O[Na])OP(=O)(O[Na])OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)n3cnc4C(=O)NC(=Nc34)N)[C@@H](O)C(O)[C@H]1O |
| IUPAC Name | [[[(2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-sodiooxyphosphoryl]oxy-[(2S,3S,5R,6R)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyphosphoryl]oxysodium |
| Molecular Weight (g/mol) | 633.305 |
| Formula Weight | 633.046 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?