missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanosine 5’-O-(2-Thiodiphosphate) Trilithium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Guanosine 5'-O-(2-Thiodiphosphate) Trilithium Salt |
| CAS | 97952-36-8 |
| Molecular Formula | C10 H12 N5 O10 P2 S . 3 Li |
| Quantity | 10 mg |
| Synonym | 5'-Guanylic acid, monoanhydride with phosphorothioic acid, trilithium salt (9CI), Trilithium GDP-βS, Guanosine 5'-(β-thiodiphosphate) trilithium salt, Guanosine 5'-O-(2-thiodiphosphate) trilithium salt, Guanosine 5'-O-(β-thiodiphosphate) trilithium salt, Guanyl-5'-yl thiophosphate trilithium salt |
| InChI Formula | InChI=1S/C10H15N5O10P2S.3Li/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(24-9)1-23-26(19,20)25-27(21,22)28;;;/h2-3,5-6,9,16-17H,1H2,(H,19,20)(H2,21,22,28)(H3,11,13,14,18);;;/q;3*+1/p-3/t3-,5-,6-,9-;;;/m1.../s1 |
| SMILES | [Li+].[Li+].[Li+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(=O)([O-])OP(=S)([O-])[O-])[C@@H](O)[C@H]3O)C(=O)N1 |
| IUPAC Name | trilithium;[(2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dioxidophosphinothioyl phosphate |
| Molecular Weight (g/mol) | 477.065 |
| Formula Weight | 477.026 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?