missing translation for 'onlineSavingsMsg'
Learn More

Guanosine 5’-O-(2-Thiodiphosphate) Trilithium Salt, TRC

Product Code. 30391618
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391618 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391618 Supplier Toronto Research Chemicals Supplier No. TRCG8387085MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Guanosine 5'-O-(2-Thiodiphosphate) Trilithium Salt
CAS 97952-36-8
Molecular Formula C10 H12 N5 O10 P2 S . 3 Li
Quantity 5 mg
Synonym 5'-Guanylic acid, monoanhydride with phosphorothioic acid, trilithium salt (9CI), Trilithium GDP-βS, Guanosine 5'-(β-thiodiphosphate) trilithium salt, Guanosine 5'-O-(2-thiodiphosphate) trilithium salt, Guanosine 5'-O-(β-thiodiphosphate) trilithium salt, Guanyl-5'-yl thiophosphate trilithium salt
InChI Formula InChI=1S/C10H15N5O10P2S.3Li/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(24-9)1-23-26(19,20)25-27(21,22)28;;;/h2-3,5-6,9,16-17H,1H2,(H,19,20)(H2,21,22,28)(H3,11,13,14,18);;;/q;3*+1/p-3/t3-,5-,6-,9-;;;/m1.../s1
SMILES [Li+].[Li+].[Li+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(=O)([O-])OP(=S)([O-])[O-])[C@@H](O)[C@H]3O)C(=O)N1
IUPAC Name trilithium;[(2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dioxidophosphinothioyl phosphate
Molecular Weight (g/mol) 477.065
Formula Weight 477.026
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.