missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanosine 5'-Diphosphate, TRC
Specifications
Specifications
| Chemical Name or Material | Guanosine 5'-Diphosphate |
| CAS | 146-91-8 |
| Molecular Formula | C10 H15 N5 O11 P2 |
| Quantity | 100 mg |
| Synonym | Guanosine 5'-(trihydrogen diphosphate) (9CI, ACI), Guanosine 5'-(trihydrogen pyrophosphate) (8CI), Guanosine pyrophosphate (6CI, 7CI), 5'-GDP, GDP, Guanosine 5'-diphosphate, Guanosine 5'-pyrophosphate, Guanosine diphosphate, Guanosine, mono(trihydrogen di |
| InChI Formula | InChI=1S/C10H15N5O11P2/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(25-9)1-24-28(22,23)26-27(19,20)21/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H2,19,20,21)(H3,11,13,14,18)/t3-,5-,6-,9-/m1/s1 |
| SMILES | NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(=O)(O)OP(=O)(O)O)[C@@H](O)[C@H]3O)C(=O)N1 |
| IUPAC Name | [(2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate |
| Molecular Weight (g/mol) | 443.2 |
| Formula Weight | 443.024 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?