missing translation for 'onlineSavingsMsg'
Learn More

Guanosine 5'-Diphosphate, TRC

Product Code. 30393191
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393191 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393191 Supplier Toronto Research Chemicals Supplier No. TRCG211380250MG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Guanosine 5'-Diphosphate
CAS 146-91-8
Molecular Formula C10 H15 N5 O11 P2
Quantity 250 mg
Synonym Guanosine 5'-(trihydrogen diphosphate) (9CI, ACI), Guanosine 5'-(trihydrogen pyrophosphate) (8CI), Guanosine pyrophosphate (6CI, 7CI), 5'-GDP, GDP, Guanosine 5'-diphosphate, Guanosine 5'-pyrophosphate, Guanosine diphosphate, Guanosine, mono(trihydrogen di
InChI Formula InChI=1S/C10H15N5O11P2/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(25-9)1-24-28(22,23)26-27(19,20)21/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H2,19,20,21)(H3,11,13,14,18)/t3-,5-,6-,9-/m1/s1
SMILES NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(=O)(O)OP(=O)(O)O)[C@@H](O)[C@H]3O)C(=O)N1
IUPAC Name [(2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate
Molecular Weight (g/mol) 443.2
Formula Weight 443.024
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.