missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Guanosine 5'-diphosphate disodium salt

Product Code. 16481027
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481027 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16481027 Supplier MedChemExpress Supplier No. HY113066A
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Guanosine 5'-diphosphate (GDP) disodium salt is a nucleoside diphosphate that activates adenosine 5'-triphosphate-sensitive K+ channel. Guanosine 5'-diphosphate disodium salt is a potential iron mobilizer, which prevents the hepcidin-ferroportin interaction and modulates the interleukin-6 (IL-6)/stat-3 pathway. Guanosine 5'-diphosphate disodium salt can be used in the research of inflammation, such as anemia of inflammation (AI).

Specifications

Molecular Weight (g/mol) 487.16 g/mol
Product Type Reference compound
Content And Storage -20°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Shipping Condition Room temperature
Quantity 50 mg
Solubility Information H2O : 125 mg/mL (256.59 mM∣Need ultrasonic) ∣DMSO : <1 mg/mL (ultrasonic∣warming∣heat to 80°C) (insoluble or slightly soluble)
For Use With (Application) COVID-19-immunoregulation
CAS 7415-69-2
Synonym GDP disodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.