missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanosine 3’,5’-Cyclic Monophosphate Sodium Salt, TRC
Spezifikation
Spezifikation
| Chemical Name or Material | Guanosine 3',5'-Cyclic Monophosphate Sodium Salt |
| CAS | 40732-48-7 |
| Molecular Formula | C10 H11 N5 O7 P . Na |
| Quantity | 100 mg |
| Synonym | Guanosine, cyclic 3',5'-(hydrogen phosphate), sodium salt (1:1), Guanosine, cyclic 3',5'-(hydrogen phosphate), monosodium salt (9CI), 4H-Furo[3,2-d]-1,3,2-dioxaphosphorin, guanosine deriv., Cyclic 3',5'-guanosine monophosphate monosodium salt, Guanosine monophosphate monosodium salt, Monosodium-GMP, Guanosine 3',5'-cyclic monophosphate sodium salt |
| InChI Formula | InChI=1S/C10H12N5O7P.Na/c11-10-13-7-4(8(17)14-10)12-2-15(7)9-5(16)6-3(21-9)1-20-23(18,19)22-6;/h2-3,5-6,9,16H,1H2,(H,18,19)(H3,11,13,14,17);/q;+1/p-1/t3-,5-,6-,9-;/m1./s1 |
| SMILES | [Na+].NC1=Nc2c(ncn2[C@@H]3O[C@@H]4COP(=O)([O-])O[C@H]4[C@H]3O)C(=O)N1 |
| IUPAC Name | sodium;9-[(4aR,6R,7R,7aS)-7-hydroxy-2-oxido-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl]-2-amino-1H-purin-6-one |
| Molecular Weight (g/mol) | 367.19 |
| Formula Weight | 367.0294 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?