missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanosine 3',5'-Cyclic Monophosphate-13C5 Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Guanosine 3',5'-Cyclic Monophosphate-13C5 Sodium Salt |
| Purity Grade Notes | HPLC |
| Molecular Formula | C513C5H11N5NaO7P |
| Quantity | 0.25 mg |
| Synonym | Guanosine Cyclic 3',5'-(Hydrogen phosphate)-13C5 Sodium Salt, Cyclic 3',5'-Guanosine Monophosphate-13C5 Monosodium Salt, Guanosine Monophosphate-13C5 Monosodium Salt, Monosodium-GMP-13C5 |
| InChI Formula | InChI=1S/C10H12N5O7P.Na/c11-10-13-7-4(8(17)14-10)12-2-15(7)9-5(16)6-3(21-9)1-20-23(18,19)22-6;/h2-3,5-6,9,16H,1H2,(H,18,19)(H3,11,13,14,17);/q;+1/p-1/t3-,5-,6-,9-;/m1./s1/i1+1,3+1,5+1,6+1,9+1; |
| SMILES | O=P1(O[Na])O[13CH2][13C@@H]2[13C@](O1)([H])[13C@@H](O)[13C@H](N(C=N3)C4=C3C(NC(N)=N4)=O)O2 |
| Molecular Weight (g/mol) | 372.15 |
| Percent Purity | >95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?