missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanosine-1'-13C Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Guanosine-1'-13C Hydrate |
| Molecular Formula | C913CH13N5O5 xH2O |
| Quantity | 1 mg |
| Synonym | 2-Amino-1,9-dihydro-9-β-D-ribofuranosyl-6H-purin-6-one-1'-13C Monohydrate, DL-Guanosine-1'-13C Monohydrate, Guanine Ribonucleoside-1'-13C Monohydrate, 9-β-D-Ribofuranosylguanine-1'-13C Monohydrate, 2-Aminoinosine-1'-13C Monohydrate, NSC 19994-1'-13C Monohydrate, Vernine-1'-13C Monohydrate |
| SMILES | O[C@@H]1[C@H](O)[C@@H](CO)O[13C@H]1N(C=N2)C3=C2C(NC(N)=N3)=O |
| Molecular Weight (g/mol) | 284.23 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?